Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033581.3 | NP_291059.1 | 1 | 1793 | |
GenBank mRNA | AK086361.1 | - | 1 | 1427 | |
AK089310.1 | - | 1 | 1840 | ||
AY013811.1 | - | 1 | 1655 | ||
BC040826.1 | - | 1 | 947 | ||
BC059175.1 | - | 1 | 1793 | 5704674 | |
BC098457.1 | - | 2 | 695 | 30653042 |