Gene Transcripts
- Gene Symbol
- Slc22a13
- Entrez Gene ID
- 102570
- Gene Name
- solute carrier family 22 (organic cation transporter), member 13
- Gene Aliases
- AI648912, OCTL1, OCTL3, ORCTL3
- Chromosome Location
- Chr. 9: 119192968 - 119209105 on Build GRCm38
- UniGene ID
- Mm.38775
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC021449.1 |
- |
3 |
652 |
|
View Full Product Details