Gene Transcripts

Gene Symbol
Oaf
Entrez Gene ID
102644
Gene Name
OAF homolog (Drosophila)
Gene Aliases
AL024221, D130038B21Rik, D9Ucla1
Chromosome Location
Chr. 9: 43221278 - 43239816 on Build GRCm38
UniGene ID
Mm.246479
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178644.3 NP_848759.1 4 1039
GenBank mRNA AK051349.1 - 4 1039
AK150279.1 - 4 1026
BC025514.1 - 4 1047 5309953
BC026022.1 - 4 1003 5098241
BC028824.1 - 4 1074 5097873
BC034836.1 - 4 1005 5125217

View Full Product Details