Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000274.3 | NP_000265.1 | 5 | 704 | |
NM_001171814.1 | NP_001165285.1 | 4 | 476 | ||
NM_001322965.1 | NP_001309894.1 | 5 | 699 | ||
NM_001322966.1 | NP_001309895.1 | 6 | 1019 | ||
NM_001322967.1 | NP_001309896.1 | 6 | 824 | ||
NM_001322968.1 | NP_001309897.1 | 7 | 911 | ||
NM_001322969.1 | NP_001309898.1 | 6 | 791 | ||
NM_001322970.1 | NP_001309899.1 | 7 | 1007 | ||
NM_001322971.1 | NP_001309900.1 | 3 | 383 | ||
NM_001322974.1 | NP_001309903.1 | 5 | 590 | ||
XM_017016279.1 | XP_016871768.1 | 5 | 2499 | ||
GenBank mRNA | AB208817.1 | - | 5 | 2489 | |
AK296032.1 | - | 4 | 423 | ||
AK312561.1 | - | 5 | 651 | ||
AK315947.1 | - | 4 | 423 | ||
BC000964.2 | - | 5 | 632 | 3449883 | |
BC016928.1 | - | 5 | 636 | 3913352 | |
CD683561.1 | - | 3 | 352 | ||
CR457045.1 | - | 4 | 571 | ||
CR749808.1 | - | 4 | 426 | ||
M12267.1 | - | 5 | 625 | ||
M14963.1 | - | 4 | 615 | ||
M23204.1 | - | 5 | 622 | ||
Y07511.1 | - | 4 | 611 |