Gene Transcripts

Gene Symbol
Tecpr2
Entrez Gene ID
104859
Gene Name
tectonin beta-propeller repeat containing 2
Gene Aliases
4930573I19Rik, AI604920, mKIAA0297
Chromosome Location
Chr. 12: 110889259 - 110972394 on Build GRCm38
UniGene ID
Mm.35583
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081057.2 NP_001074526.1 7 1104
NM_001289510.1 NP_001276439.1 6 1052
NM_175336.3 NP_780545.2 7 1104
GenBank mRNA AK031736.1 - 6 1009
AK129113.1 - 6 1056
AK147592.1 - 6 1052
BC041778.1 - 4 696
BC138354.1 - 6 1047 8861374
BC145250.1 - 6 1047

View Full Product Details