Gene Transcripts

Gene Symbol
Arrdc3
Entrez Gene ID
105171
Gene Name
arrestin domain containing 3
Gene Aliases
AI450344, mKIAA1376
Chromosome Location
Chr. 13: 80883338 - 80896042 on Build GRCm38
UniGene ID
Mm.423137
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042591.1 NP_001036056.1 5, 6 1099
GenBank mRNA AK084951.1 - 5, 6 1110
AK088288.1 - 5, 6 1114
AK122499.1 - 5, 6 1086
AK163364.1 - 5, 6 644
BC023838.1 - 2, 3 317
BC031125.1 - 3, 4 477
BC044762.1 - 5, 6 1097
BC054844.1 - 5, 6 1099 6493441
BC098474.1 - 5, 6 1093 30094818
DQ861994.1 - 5, 6 844

View Full Product Details