Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164804.1 | NP_001158276.1 | 2 | 261 | |
NM_178893.4 | NP_849224.2 | 3 | 486 | ||
GenBank mRNA | AK030097.1 | - | 3 | 489 | |
AK030971.1 | - | 2 | 217 | ||
AK044060.1 | - | 2 | 223 | ||
AK054394.1 | - | 2 | 262 | ||
AK152490.1 | - | 2 | 250 | ||
AK153134.1 | - | 2 | 250 | ||
AK154247.1 | - | 2 | 218 | ||
AK170184.1 | - | 2 | 219 | ||
BC138731.1 | - | 2 | 286 | 8861753 | |
BC138732.1 | - | 2 | 286 | 8861754 |