Gene Transcripts

Gene Symbol
Grm5
Entrez Gene ID
108071
Gene Name
glutamate receptor, metabotropic 5
Gene Aliases
6430542K11Rik, AI850523, Glu5R, Gprc1e, mGluR5, mGluR5b
Chromosome Location
Chr. 7: 87583981 - 88135063 on Build GRCm38
UniGene ID
Mm.235018
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081414.2 NP_001074883.1 3 1350
NM_001143834.1 NP_001137306.1 3 1350
GenBank mRNA AK032422.1 - 1 178
AK136840.1 - 3 1139
BC096533.1 - 1 735
EF508472.1 - 1 632

View Full Product Details