Gene Transcripts

Gene Symbol
Ciapin1
Entrez Gene ID
109006
Gene Name
cytokine induced apoptosis inhibitor 1
Gene Aliases
2810413N20Rik, AA617265, AU021794, anamorsin
Chromosome Location
Chr. 8: 94819818 - 94838340 on Build GRCm38
UniGene ID
Mm.27582
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134141.4 NP_598902.1 2, 3 274
GenBank mRNA AK076116.1 - 2, 3 230
AK079615.1 - 2, 3 236
AK088757.1 - 1, 2 201
AK136821.1 - 2, 3 268
AK146282.1 - 2, 3 222
AK163705.1 - 2, 3 253
AK168252.1 - 2, 3 224
AK188252.1 - 1, 2 19
AK210999.1 - 1, 2 19
AY523555.1 - 1, 2 195
BC016261.1 - 1, 2 192 4920560
BC021864.1 - 2, 3 214 5123958
BC021949.1 - 1, 2 25
BY283649.1 - 2, 3 293

View Full Product Details