Gene Transcripts

Gene Symbol
Prr5
Entrez Gene ID
109270
Gene Name
proline rich 5 (renal)
Gene Aliases
AU043908, Arhgap8, C030017C09Rik, C78947, Protor-1, Protor1
Chromosome Location
Chr. 15: 84663171 - 84706409 on Build GRCm38
UniGene ID
Mm.291372
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146061.4 NP_666173.4 7 863
GenBank mRNA AF482997.1 - 7 858
AK046057.1 - 8 680
AK081231.1 - 7 859

View Full Product Details