Gene Transcripts

Gene Symbol
LINC00337
Entrez Gene ID
148645
Gene Name
long intergenic non-protein coding RNA 337
Gene Aliases
C1orf211, NCRNA00337
Chromosome Location
Chr. 1: 6236240 - 6239444 on Build GRCh38
UniGene ID
Hs.185688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_103534.1 5 1427
GenBank mRNA BC030279.1 - 2 1058

View Full Product Details