Gene Transcripts

Gene Symbol
DNASE1L3
Entrez Gene ID
1776
Gene Name
deoxyribonuclease I like 3
Gene Aliases
DHP2, DNAS1L3, LSD, SLEB16
Chromosome Location
Chr.3: 58192626 - 58211003 on Build GRCh38
Uni Gene
Hs.476453
Interrogated Sequence Translated Protein
RefSeq NM_001256560.1 NP_001243489.1
NM_004944.3 NP_004935.1
GenBank mRNA AB209613.1 -
AF047354.1 -
AI591302.1 -
AI660448.1 -
AK131323.1 -
AK301263.1 -
AK313303.1 -
BC015831.1 -
BI521582.1 -
BI834035.1 -
DB164811.1 -
ENST00000394549.0 -
U56814.1 -

View Full Product Details