Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001256560.1 | NP_001243489.1 |
NM_004944.3 | NP_004935.1 | |
GenBank mRNA | AB209613.1 | - |
AF047354.1 | - | |
AI591302.1 | - | |
AI660448.1 | - | |
AK131323.1 | - | |
AK301263.1 | - | |
AK313303.1 | - | |
BC015831.1 | - | |
BI521582.1 | - | |
BI834035.1 | - | |
DB164811.1 | - | |
ENST00000394549.0 | - | |
U56814.1 | - |