Gene Transcripts

Gene Symbol
Lsd-2
Entrez Gene ID
32437
Gene Name
Lipid storage droplet-2
Gene Aliases
145098_at, CG9057, DmPLIN2, Dmel\CG9057, Dmel_CG9057, LSD, LSD-2, LSD2, LSP-2, Lsd2, PLIN2, Plin2, dmPLIN2, lsd-2, lsd2, plin2
Chromosome Location
Chr.X: 15069127 - 15076220 on Build Release_6_plus_ISO1_MT
Uni Gene
Dm.4079
Species Specific ID (Flybase ID)
FBgn0030608
Interrogated Sequence Translated Protein
RefSeq NM_001042811.2 NP_001036276.1
NM_001042812.2 NP_001036277.2
NM_001298332.1 NP_001285261.1
NM_001298333.1 NP_001285262.1
NM_132768.3 NP_572996.1
GenBank mRNA AY060242.1 -

View Full Product Details