Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008768160.1 | XP_008766382.1 | 6 | 699 | |
XM_008768161.1 | XP_008766383.1 | 6 | 938 | ||
XM_008768162.1 | XP_008766384.1 | 6 | 699 | ||
XM_008768163.1 | XP_008766385.1 | 6 | 699 | ||
XM_008768164.1 | XP_008766386.1 | 5 | 802 |