Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 1393875-1394366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC6A3
Entrez Gene ID
6531
Gene Name
solute carrier family 6 member 3
Gene Aliases
DAT, DAT1, PKDYS
UniGene
Hs.406
Gene Chromosome Location
Chr.5: 1392790-1445428 on Build GRCh38
Transcripts
NM_001044.4

SNP Detail

SNP ID Chromosome Location
rs907322448 Chr.5: 1394198-1394198 on Build GRCh38
rs933759226 Chr.5: 1394330-1394330 on Build GRCh38
rs185128091 Chr.5: 1394181-1394181 on Build GRCh38
rs1052864071 Chr.5: 1394005-1394005 on Build GRCh38
rs564054633 Chr.5: 1394329-1394329 on Build GRCh38
rs973500193 Chr.5: 1393939-1393939 on Build GRCh38
rs563207652 Chr.5: 1393926-1393926 on Build GRCh38
rs534365130 Chr.5: 1394001-1394001 on Build GRCh38
rs1004971327 Chr.5: 1393919-1393919 on Build GRCh38
rs985325406 Chr.5: 1393923-1393923 on Build GRCh38
rs953725938 Chr.5: 1394123-1394123 on Build GRCh38
rs900139632 Chr.5: 1394129-1394129 on Build GRCh38
rs557156287 Chr.5: 1394113-1394113 on Build GRCh38
rs573168726 Chr.5: 1394170-1394170 on Build GRCh38
rs990162691 Chr.5: 1394229-1394229 on Build GRCh38
rs535619847 Chr.5: 1394039-1394039 on Build GRCh38
rs923712420 Chr.5: 1394104-1394104 on Build GRCh38
rs977608827 Chr.5: 1394089-1394089 on Build GRCh38
rs539912214 Chr.5: 1394132-1394132 on Build GRCh38
rs545881658 Chr.5: 1394328-1394328 on Build GRCh38
rs951937530 Chr.5: 1393897-1393897 on Build GRCh38
rs997817665 Chr.5: 1393901-1393901 on Build GRCh38
rs981437749 Chr.5: 1394002-1394002 on Build GRCh38
rs45623739 Chr.5: 1394185-1394185 on Build GRCh38
rs997870126 Chr.5: 1394047-1394047 on Build GRCh38
rs919331997 Chr.5: 1393962-1393962 on Build GRCh38
rs1000362262 Chr.5: 1394004-1394004 on Build GRCh38
rs962559079 Chr.5: 1393930-1393930 on Build GRCh38
rs773058784 Chr.5: 1393992-1393992 on Build GRCh38
rs145747191 Chr.5: 1394182-1394182 on Build GRCh38
rs200979189 Chr.5: 1393899-1393899 on Build GRCh38
rs917266106 Chr.5: 1394241-1394241 on Build GRCh38
rs189117846 Chr.5: 1394200-1394200 on Build GRCh38
rs990335930 Chr.5: 1394033-1394033 on Build GRCh38
rs936157398 Chr.5: 1394305-1394305 on Build GRCh38
rs921600468 Chr.5: 1394326-1394326 on Build GRCh38
rs958967861 Chr.5: 1394032-1394032 on Build GRCh38
rs147961632 Chr.5: 1393963-1393963 on Build GRCh38
rs141895758 Chr.5: 1394010-1394010 on Build GRCh38
rs373686040 Chr.5: 1394223-1394223 on Build GRCh38
rs534546248 Chr.5: 1394015-1394015 on Build GRCh38
rs931892834 Chr.5: 1394146-1394147 on Build GRCh38
rs948955618 Chr.5: 1393971-1393971 on Build GRCh38
rs1016851408 Chr.5: 1394169-1394169 on Build GRCh38
rs28363168 Chr.5: 1394274-1394274 on Build GRCh38
rs944451640 Chr.5: 1394307-1394307 on Build GRCh38
rs193282661 Chr.5: 1393993-1393993 on Build GRCh38
rs1057020098 Chr.5: 1393966-1393966 on Build GRCh38
rs950778497 Chr.5: 1394203-1394203 on Build GRCh38
rs891566029 Chr.5: 1394327-1394327 on Build GRCh38
rs926193031 Chr.5: 1394298-1394298 on Build GRCh38
rs948730728 Chr.5: 1394245-1394245 on Build GRCh38
rs958666990 Chr.5: 1394205-1394205 on Build GRCh38
rs558015840 Chr.5: 1394151-1394151 on Build GRCh38
rs138415711 Chr.5: 1394325-1394325 on Build GRCh38
rs541997406 Chr.5: 1394222-1394222 on Build GRCh38
rs914746542 Chr.5: 1394034-1394034 on Build GRCh38
rs747708228 Chr.5: 1393927-1393927 on Build GRCh38
rs1007244969 Chr.5: 1394146-1394146 on Build GRCh38
rs2652504 Chr.5: 1394006-1394006 on Build GRCh38
rs1027501845 Chr.5: 1393896-1393896 on Build GRCh38
rs1029584610 Chr.5: 1393912-1393912 on Build GRCh38
rs1034329089 Chr.5: 1394201-1394201 on Build GRCh38
rs917524919 Chr.5: 1393967-1393967 on Build GRCh38
rs188668123 Chr.5: 1393914-1393914 on Build GRCh38
rs374336620 Chr.5: 1393940-1393940 on Build GRCh38
rs192107796 Chr.5: 1394206-1394206 on Build GRCh38
rs28363169 Chr.5: 1394119-1394119 on Build GRCh38
rs992854950 Chr.5: 1394215-1394215 on Build GRCh38
rs931675156 Chr.5: 1394344-1394344 on Build GRCh38

View Full Product Details