Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 640164-640667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRD4
Entrez Gene ID
1815
Gene Name
dopamine receptor D4
Gene Aliases
D4DR
UniGene
Hs.99922
Gene Chromosome Location
Chr.11: 637305-640706 on Build GRCh38
Transcripts
NM_000797.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5473187 c.998G>A p.R333Q Chr.11: 640247-640247 on Build GRCh38
2163289 c.1088T>A p.F363Y Chr.11: 640431-640431 on Build GRCh38
4035104 c.1254C>G p.C418W Chr.11: 640597-640597 on Build GRCh38
6649284 c.1080G>A p.T360T Chr.11: 640423-640423 on Build GRCh38
7348815 c.1129G>A p.V377M Chr.11: 640472-640472 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754680843 Chr.11: 640508-640508 on Build GRCh38
rs753583846 Chr.11: 640552-640553 on Build GRCh38
rs540217661 Chr.11: 640212-640212 on Build GRCh38
rs777913224 Chr.11: 640523-640523 on Build GRCh38
rs779255532 Chr.11: 640584-640584 on Build GRCh38
rs758250713 Chr.11: 640279-640279 on Build GRCh38
rs778516673 Chr.11: 640448-640448 on Build GRCh38
rs757838518 Chr.11: 640521-640521 on Build GRCh38
rs780276066 Chr.11: 640312-640312 on Build GRCh38
rs769472877 Chr.11: 640582-640582 on Build GRCh38
rs772240299 Chr.11: 640585-640585 on Build GRCh38
rs773248526 Chr.11: 640471-640471 on Build GRCh38
rs370350460 Chr.11: 640489-640489 on Build GRCh38
rs770293048 Chr.11: 640644-640644 on Build GRCh38
rs767356732 Chr.11: 640373-640373 on Build GRCh38
rs763081988 Chr.11: 640212-640215 on Build GRCh38
rs773219389 Chr.11: 640546-640546 on Build GRCh38
rs748926835 Chr.11: 640445-640445 on Build GRCh38
rs567735616 Chr.11: 640341-640341 on Build GRCh38
rs781541553 Chr.11: 640348-640349 on Build GRCh38
rs942969082 Chr.11: 640225-640225 on Build GRCh38
rs769459852 Chr.11: 640399-640399 on Build GRCh38
rs749376166 Chr.11: 640319-640319 on Build GRCh38
rs930253839 Chr.11: 640544-640544 on Build GRCh38
rs757701471 Chr.11: 640621-640622 on Build GRCh38
rs969409206 Chr.11: 640424-640425 on Build GRCh38
rs368379171 Chr.11: 640393-640393 on Build GRCh38
rs761647489 Chr.11: 640346-640346 on Build GRCh38
rs756530079 Chr.11: 640499-640499 on Build GRCh38
rs775547278 Chr.11: 640209-640209 on Build GRCh38
rs529779269 Chr.11: 640204-640204 on Build GRCh38
rs766820146 Chr.11: 640555-640555 on Build GRCh38
rs749061233 Chr.11: 640622-640622 on Build GRCh38
rs772127667 Chr.11: 640545-640545 on Build GRCh38
rs370568420 Chr.11: 640576-640576 on Build GRCh38
rs977842521 Chr.11: 640201-640201 on Build GRCh38
rs370427849 Chr.11: 640602-640602 on Build GRCh38
rs750314769 Chr.11: 640614-640614 on Build GRCh38
rs1034789684 Chr.11: 640273-640273 on Build GRCh38
rs994258425 Chr.11: 640247-640247 on Build GRCh38
rs758008715 Chr.11: 640566-640566 on Build GRCh38
rs779660727 Chr.11: 640400-640400 on Build GRCh38
rs74991942 Chr.11: 640502-640502 on Build GRCh38
rs748120920 Chr.11: 640409-640409 on Build GRCh38
rs773300863 Chr.11: 640586-640586 on Build GRCh38
rs757652339 Chr.11: 640396-640396 on Build GRCh38
rs372805272 Chr.11: 640243-640243 on Build GRCh38
rs781710797 Chr.11: 640622-640622 on Build GRCh38
rs12720410 Chr.11: 640254-640254 on Build GRCh38
rs1870724 Chr.11: 640369-640369 on Build GRCh38
rs759787985 Chr.11: 640558-640558 on Build GRCh38
rs936434870 Chr.11: 640314-640314 on Build GRCh38
rs377281442 Chr.11: 640565-640565 on Build GRCh38
rs976890423 Chr.11: 640635-640635 on Build GRCh38
rs749338488 Chr.11: 640355-640362 on Build GRCh38
rs200635197 Chr.11: 640378-640378 on Build GRCh38
rs989478549 Chr.11: 640237-640237 on Build GRCh38
rs963856946 Chr.11: 640629-640629 on Build GRCh38
rs773478489 Chr.11: 640339-640339 on Build GRCh38
rs750927378 Chr.11: 640498-640498 on Build GRCh38
rs748678312 Chr.11: 640329-640329 on Build GRCh38
rs199628904 Chr.11: 640411-640411 on Build GRCh38
rs373483649 Chr.11: 640577-640577 on Build GRCh38
rs201770727 Chr.11: 640496-640496 on Build GRCh38
rs139758515 Chr.11: 640534-640534 on Build GRCh38
rs770929160 Chr.11: 640526-640526 on Build GRCh38
rs889445416 Chr.11: 640221-640221 on Build GRCh38
rs772094160 Chr.11: 640456-640456 on Build GRCh38
rs200086731 Chr.11: 640418-640418 on Build GRCh38
rs780770081 Chr.11: 640353-640353 on Build GRCh38
rs948002154 Chr.11: 640240-640240 on Build GRCh38
rs767234596 Chr.11: 640329-640333 on Build GRCh38
rs760603804 Chr.11: 640426-640428 on Build GRCh38
rs1038073887 Chr.11: 640302-640302 on Build GRCh38
rs745604469 Chr.11: 640533-640533 on Build GRCh38
rs571849183 Chr.11: 640250-640250 on Build GRCh38
rs763243971 Chr.11: 640483-640483 on Build GRCh38
rs949193977 Chr.11: 640194-640194 on Build GRCh38
rs754493615 Chr.11: 640342-640342 on Build GRCh38
rs34773603 Chr.11: 640450-640450 on Build GRCh38
rs201902931 Chr.11: 640607-640607 on Build GRCh38
rs776711338 Chr.11: 640634-640634 on Build GRCh38
rs564667137 Chr.11: 640341-640342 on Build GRCh38
rs373242165 Chr.11: 640537-640537 on Build GRCh38
rs571854363 Chr.11: 640274-640274 on Build GRCh38
rs867503103 Chr.11: 640457-640457 on Build GRCh38
rs982704550 Chr.11: 640447-640447 on Build GRCh38
rs142719624 Chr.11: 640561-640561 on Build GRCh38
rs781166730 Chr.11: 640528-640528 on Build GRCh38
rs936338365 Chr.11: 640209-640209 on Build GRCh38
rs769799392 Chr.11: 640205-640206 on Build GRCh38
rs149408856 Chr.11: 640497-640497 on Build GRCh38
rs763179251 Chr.11: 640423-640423 on Build GRCh38
rs772638664 Chr.11: 640332-640332 on Build GRCh38
rs754464416 Chr.11: 640570-640571 on Build GRCh38
rs758282163 Chr.11: 640480-640480 on Build GRCh38
rs768647266 Chr.11: 640368-640369 on Build GRCh38
rs766200986 Chr.11: 640341-640341 on Build GRCh38
rs115232659 Chr.11: 640578-640578 on Build GRCh38
rs776995219 Chr.11: 640601-640601 on Build GRCh38
rs370728840 Chr.11: 640617-640617 on Build GRCh38
rs958733560 Chr.11: 640275-640275 on Build GRCh38
rs544372546 Chr.11: 640453-640453 on Build GRCh38
rs12720413 Chr.11: 640370-640370 on Build GRCh38
rs1047301979 Chr.11: 640548-640548 on Build GRCh38
rs750837353 Chr.11: 640437-640437 on Build GRCh38
rs778407682 Chr.11: 640513-640513 on Build GRCh38
rs745538181 Chr.11: 640354-640354 on Build GRCh38
rs759768563 Chr.11: 640194-640201 on Build GRCh38
rs773136049 Chr.11: 640420-640421 on Build GRCh38
rs761666406 Chr.11: 640368-640368 on Build GRCh38
rs763981192 Chr.11: 640345-640345 on Build GRCh38
rs377409215 Chr.11: 640590-640590 on Build GRCh38
rs766013337 Chr.11: 640238-640238 on Build GRCh38
rs369869458 Chr.11: 640606-640606 on Build GRCh38
rs755832118 Chr.11: 640337-640339 on Build GRCh38
rs746193982 Chr.11: 640398-640398 on Build GRCh38
rs746302362 Chr.11: 640349-640350 on Build GRCh38
rs200996360 Chr.11: 640253-640253 on Build GRCh38
rs769372047 Chr.11: 640358-640358 on Build GRCh38
rs762019616 Chr.11: 640539-640539 on Build GRCh38
rs886039342 Chr.11: 640417-640418 on Build GRCh38
rs923696945 Chr.11: 640309-640309 on Build GRCh38
rs537286059 Chr.11: 640351-640351 on Build GRCh38
rs773162405 Chr.11: 640412-640412 on Build GRCh38
rs760248377 Chr.11: 640216-640216 on Build GRCh38
rs752870951 Chr.11: 640380-640380 on Build GRCh38
rs1870723 Chr.11: 640349-640349 on Build GRCh38
rs781259520 Chr.11: 640612-640612 on Build GRCh38
rs936463 Chr.11: 640432-640432 on Build GRCh38
rs770534087 Chr.11: 640342-640342 on Build GRCh38
rs765195080 Chr.11: 640264-640264 on Build GRCh38
rs201878429 Chr.11: 640305-640305 on Build GRCh38
rs377630101 Chr.11: 640616-640616 on Build GRCh38
rs752038369 Chr.11: 640612-640621 on Build GRCh38
rs761199673 Chr.11: 640550-640550 on Build GRCh38
rs779620136 Chr.11: 640328-640328 on Build GRCh38
rs775015409 Chr.11: 640359-640359 on Build GRCh38
rs750118457 Chr.11: 640375-640375 on Build GRCh38
rs200686422 Chr.11: 640187-640187 on Build GRCh38
rs779136633 Chr.11: 640348-640348 on Build GRCh38
rs199815231 Chr.11: 640265-640265 on Build GRCh38
rs888502358 Chr.11: 640258-640258 on Build GRCh38
rs773676450 Chr.11: 640408-640408 on Build GRCh38
rs764250733 Chr.11: 640426-640426 on Build GRCh38
rs747730168 Chr.11: 640519-640519 on Build GRCh38
rs779748850 Chr.11: 640441-640441 on Build GRCh38
rs910939150 Chr.11: 640529-640529 on Build GRCh38
rs755079455 Chr.11: 640326-640326 on Build GRCh38
rs750035392 Chr.11: 640331-640332 on Build GRCh38
rs748836831 Chr.11: 640405-640405 on Build GRCh38
rs757816571 Chr.11: 640433-640433 on Build GRCh38
rs774197789 Chr.11: 640322-640336 on Build GRCh38
rs12720411 Chr.11: 640350-640350 on Build GRCh38
rs780003868 Chr.11: 640183-640183 on Build GRCh38
rs756441938 Chr.11: 640438-640438 on Build GRCh38
rs908434388 Chr.11: 640492-640492 on Build GRCh38
rs374911834 Chr.11: 640472-640472 on Build GRCh38
rs747346759 Chr.11: 640340-640340 on Build GRCh38
rs776312356 Chr.11: 640343-640343 on Build GRCh38
rs761626955 Chr.11: 640325-640363 on Build GRCh38
rs765531496 Chr.11: 640604-640604 on Build GRCh38
rs774515243 Chr.11: 640494-640494 on Build GRCh38
rs200069417 Chr.11: 640281-640281 on Build GRCh38
rs770833402 Chr.11: 640475-640475 on Build GRCh38
rs201086345 Chr.11: 640344-640344 on Build GRCh38
rs774977981 Chr.11: 640202-640202 on Build GRCh38
rs761777086 Chr.11: 640495-640495 on Build GRCh38
rs749350638 Chr.11: 640185-640185 on Build GRCh38
rs531147000 Chr.11: 640623-640623 on Build GRCh38
rs778207427 Chr.11: 640390-640390 on Build GRCh38
rs766106162 Chr.11: 640459-640460 on Build GRCh38
rs770981157 Chr.11: 640628-640628 on Build GRCh38
rs752951790 Chr.11: 640530-640530 on Build GRCh38
rs989509727 Chr.11: 640343-640344 on Build GRCh38
rs775742671 Chr.11: 640645-640645 on Build GRCh38
rs746953417 Chr.11: 640524-640524 on Build GRCh38
rs754278373 Chr.11: 640557-640557 on Build GRCh38
rs867464843 Chr.11: 640467-640467 on Build GRCh38
rs781624855 Chr.11: 640397-640397 on Build GRCh38
rs757585485 Chr.11: 640344-640345 on Build GRCh38
rs1035456517 Chr.11: 640605-640605 on Build GRCh38
rs746363936 Chr.11: 640638-640638 on Build GRCh38
rs764187592 Chr.11: 640488-640488 on Build GRCh38
rs774292889 Chr.11: 640367-640368 on Build GRCh38
rs767270843 Chr.11: 640385-640386 on Build GRCh38
rs756322987 Chr.11: 640307-640307 on Build GRCh38
rs754520069 Chr.11: 640446-640446 on Build GRCh38
rs758519223 Chr.11: 640387-640387 on Build GRCh38
rs201886833 Chr.11: 640417-640417 on Build GRCh38
rs748588687 Chr.11: 640203-640203 on Build GRCh38
rs552947350 Chr.11: 640347-640347 on Build GRCh38
rs970549690 Chr.11: 640295-640295 on Build GRCh38
rs778304400 Chr.11: 640342-640344 on Build GRCh38
rs764073725 Chr.11: 640293-640302 on Build GRCh38
rs577492707 Chr.11: 640500-640500 on Build GRCh38
rs776865230 Chr.11: 640190-640190 on Build GRCh38
rs772551041 Chr.11: 640205-640205 on Build GRCh38
rs764668307 Chr.11: 640597-640598 on Build GRCh38
rs184197348 Chr.11: 640589-640589 on Build GRCh38

View Full Product Details