Gene Transcripts

Gene Symbol
LOC101725754
Entrez Gene ID
101725754
Gene Name
cocaine esterase
Gene Aliases
Ces2, GW7_12028
Chromosome Location
Chr.Un|NW_004624746: 19357304 - 19366345 on Build HetGla_female_1.0
Interrogated Sequence Translated Protein
RefSeq XM_004843076.3 XP_004843133.1
XM_004843077.3 XP_004843134.1
XM_013071580.2 XP_012927034.1

View Full Product Details