Gene Transcripts

Gene Symbol
Sycn
Entrez Gene ID
68416
Gene Name
syncollin
Gene Aliases
0910001K16Rik, 1810038B08Rik, SYL
Chromosome Location
Chr. 7: 28540885 - 28542210 on Build GRCm38
UniGene ID
Mm.25210
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026716.3 NP_080992.3 1 167
GenBank mRNA AK003083.1 - 1 175
BC019567.1 - 1 167 4208065

View Full Product Details