Gene Transcripts

Gene Symbol
MAP3K6
Entrez Gene ID
9064
Gene Name
mitogen-activated protein kinase kinase kinase 6
Gene Aliases
ASK2, MAPKKK6, MEKK6
Chromosome Location
Chr. 1: 27355179 - 27368037 on Build GRCh38
UniGene ID
Hs.194694
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001297609.1 NP_001284538.1 18, 19 2836
NM_004672.4 NP_004663.3 19, 20 2860
XM_017002771.1 XP_016858260.1 17, 18 2241
GenBank mRNA AB167411.1 - 19, 20 2591
AB208805.1 - 18, 19 2305
AF100318.1 - 16, 17 2005
BC015914.1 - 15, 16 1762
BC109032.2 - 10, 11 1139
BC129950.1 - 11, 12 1290
BC129951.1 - 11, 12 1290

View Full Product Details