Gene Transcripts
- Gene Symbol
- ONECUT2
- Entrez Gene ID
- 9480
- Gene Name
- one cut homeobox 2
- Gene Aliases
- OC-2, OC2
- Chromosome Location
- Chr. 18: 57435420 - 57491298 on Build GRCh38
- UniGene ID
- Hs.194725
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004852.2 |
NP_004843.2
|
2 |
1561 |
|
GenBank mRNA |
Y18198.1 |
- |
2 |
1560 |
|
View Full Product Details