Gene Transcripts

Gene Symbol
ONECUT2
Entrez Gene ID
9480
Gene Name
one cut homeobox 2
Gene Aliases
OC-2, OC2
Chromosome Location
Chr. 18: 57435420 - 57491298 on Build GRCh38
UniGene ID
Hs.194725
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004852.2 NP_004843.2 2 1596
GenBank mRNA Y18198.1 - 2 1595

View Full Product Details