Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_020769.2 | NP_065820.1 | 3 | 2325 | |
XM_011530998.2 | XP_011529300.1 | 3 | 2347 | ||
XM_011530999.2 | XP_011529301.1 | 4 | 3196 | ||
XM_017029694.1 | XP_016885183.1 | 2 | 2255 | ||
XM_017029695.1 | XP_016885184.1 | 5 | 3329 | ||
GenBank mRNA | AB037739.1 | - | 3 | 2325 | |
AY121804.1 | - | 3 | 2325 | ||
BC113670.1 | - | 2 | 2166 | 8322722 | |
BC117184.1 | - | 2 | 2166 | 40125735 |