Gene Transcripts

Gene Symbol
CHST12
Entrez Gene ID
55501
Gene Name
carbohydrate (chondroitin 4) sulfotransferase 12
Gene Aliases
C4S-2, C4ST-2, C4ST2
Chromosome Location
Chr. 7: 2403489 - 2434607 on Build GRCh38
UniGene ID
Hs.744987
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243794.1 NP_001230723.1 2 1344
NM_001243795.1 NP_001230724.1 2 1358
NM_018641.4 NP_061111.1 2 1372
XM_011515443.2 XP_011513745.1 2 1497
XM_011515444.2 XP_011513746.1 2 1320
GenBank mRNA AF239822.1 - 2 1283
AJ289131.1 - 1 1270
AK223191.1 - 2 1293
AK223211.1 - 2 1293
AK313484.1 - 2 1293
AY358574.1 - 2 1308
BC002918.1 - 2 1325 3955396
BC015954.1 - 2 1325
BC095492.1 - 2 1332 30511863

View Full Product Details