Gene Transcripts

Gene Symbol
ADAM18
Entrez Gene ID
8749
Gene Name
ADAM metallopeptidase domain 18
Gene Aliases
ADAM27, tMDCIII
Chromosome Location
Chr. 8: 39584568 - 39730064 on Build GRCh38
UniGene ID
Hs.127930
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320313.1 NP_001307242.1 15 1656
NM_014237.2 NP_055052.1 16 1728
NR_135201.1 14 1366
GenBank mRNA AJ133004.1 - 16 1673
AK313961.1 - 16 1718
AY358321.1 - 15 1643
BC121045.1 - 16 1673 40117918
BC121046.1 - 14 1311

View Full Product Details