Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317395.1 | NP_001304324.1 | 2 | 411 | |
NM_001317399.1 | NP_001304328.1 | 2 | 411 | ||
NM_031954.4 | NP_114160.1 | 2 | 411 | ||
NR_133897.1 | 2 | 411 | |||
GenBank mRNA | AF113208.1 | - | 2 | 156 | |
AK027543.1 | - | 2 | 217 | ||
AK056912.1 | - | 2 | 1816 | ||
AK092170.1 | - | 2 | 200 | ||
AK222548.1 | - | 2 | 167 | ||
AY205299.1 | - | 2 | 217 | ||
AY597809.1 | - | 1 | 129 | ||
BC040062.1 | - | 2 | 164 | 5494016 | |
BX333975.2 | - | 2 | 161 | ||
DA440672.1 | - | 2 | 411 |