Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001190997.2 | NP_001177926.1 | 5 | 532 | |
NM_016615.4 | NP_057699.2 | 7 | 808 | ||
XM_006719008.3 | XP_006719071.1 | 2 | 595 | ||
XM_011521012.2 | XP_011519314.1 | 7 | 876 | ||
XM_011521013.1 | XP_011519315.1 | 8 | 944 | ||
XM_011521014.1 | XP_011519316.1 | 6 | 700 | ||
XM_017019842.1 | XP_016875331.1 | 2 | 601 | ||
XM_017019843.1 | XP_016875332.1 | 7 | 771 | ||
XM_017019844.1 | XP_016875333.1 | 7 | 771 | ||
XM_017019845.1 | XP_016875334.1 | 3 | 193 | ||
XM_017019846.1 | XP_016875335.1 | 7 | 771 | ||
XM_017019847.1 | XP_016875336.1 | 7 | 771 | ||
GenBank mRNA | AK295904.1 | - | 7 | 633 | |
AK296127.1 | - | 5 | 495 | ||
AK296209.1 | - | 8 | 739 | ||
AK313511.1 | - | 7 | 764 | ||
BC022392.1 | - | 7 | 771 | 4612245 | |
U76343.2 | - | 7 | 683 |