Gene Transcripts

Gene Symbol
UBA7
Entrez Gene ID
7318
Gene Name
ubiquitin like modifier activating enzyme 7
Gene Aliases
D8, UBA1B, UBE1L, UBE2
Chromosome Location
Chr. 3: 49805205 - 49813958 on Build GRCh38
UniGene ID
Hs.16695
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003335.2 NP_003326.2 4 602
XM_005265430.2 XP_005265487.1 4 586
XM_006713321.2 XP_006713384.1 4 586
XM_011534070.1 XP_011532372.1 4 586
XM_011534071.1 XP_011532373.1 4 586
XM_011534072.1 XP_011532374.1 4 586
GenBank mRNA BC006378.2 - 4 527
BT007026.1 - 4 431
L13852.1 - 4 595

View Full Product Details