Gene Transcripts
- Gene Symbol
- UBE2C
- Entrez Gene ID
- 11065
- Gene Name
- ubiquitin conjugating enzyme E2 C
- Gene Aliases
- UBCH10, dJ447F3.2
- Chromosome Location
- Chr. 20: 45812576 - 45816957 on Build GRCh38
- UniGene ID
- Hs.93002
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281741.1 |
NP_001268670.1
|
3, 4 |
411 |
|
NM_001281742.1 |
NP_001268671.1
|
3, 4 |
347 |
|
NM_007019.3 |
NP_008950.1
|
4, 5 |
552 |
|
NM_181799.2 |
NP_861515.1
|
4, 5 |
498 |
|
NM_181800.2 |
NP_861516.1
|
3, 4 |
465 |
|
NM_181801.3 |
NP_861517.1
|
4, 5 |
589 |
|
NR_104036.1 |
|
2, 3 |
260 |
|
NR_104037.1 |
|
3, 4 |
355 |
|
GenBank mRNA |
AK311949.1 |
- |
4, 5 |
519 |
|
BC007656.2 |
- |
4, 5 |
475 |
3345575 |
BC016292.1 |
- |
4, 5 |
481 |
3910286 |
BC028161.1 |
- |
3, 4 |
1091 |
|
BC050736.1 |
- |
4, 5 |
502 |
6503915 |
BI858659.1 |
- |
4, 5 |
589 |
|
BM556795.1 |
- |
4, 5 |
416 |
|
BT007300.1 |
- |
4, 5 |
432 |
|
BT020165.1 |
- |
4, 5 |
432 |
|
BT020166.1 |
- |
4, 5 |
432 |
|
BU169315.1 |
- |
4, 5 |
439 |
|
BU171488.1 |
- |
3, 4 |
371 |
|
BU844974.1 |
- |
3, 4 |
367 |
|
CB143441.1 |
- |
2, 3 |
178 |
|
CK904708.1 |
- |
3, 4 |
377 |
|
CN354813.1 |
- |
3, 4 |
248 |
|
CN354815.1 |
- |
3, 4 |
290 |
|
CR456988.1 |
- |
4, 5 |
432 |
|
U73379.1 |
- |
4, 5 |
472 |
|
View Full Product Details