Gene Transcripts

Gene Symbol
TULP1
Entrez Gene ID
7287
Gene Name
tubby like protein 1
Gene Aliases
LCA15, RP14, TUBL1
Chromosome Location
Chr. 6: 35497874 - 35512902 on Build GRCh38
UniGene ID
Hs.485208
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289395.1 NP_001276324.1 4 283
NM_003322.4 NP_003313.3 5 442
GenBank mRNA AB593123.1 - 5 442
AB593124.1 - 5 428
BC032714.1 - 4 268 5495136
BC065261.1 - 4 251 6144003
U82468.1 - 5 434

View Full Product Details