Gene Transcripts
- Gene Symbol
- KCTD3
- Entrez Gene ID
- 51133
- Gene Name
- potassium channel tetramerization domain containing 3
- Gene Aliases
- NY-REN-45
- Chromosome Location
- Chr. 1: 215567185 - 215621821 on Build GRCh38
- UniGene ID
- Hs.335139
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001319294.1 |
NP_001306223.1
|
6, 7 |
709 |
|
NM_001319295.1 |
NP_001306224.1
|
6, 7 |
781 |
|
NM_016121.4 |
NP_057205.2
|
6, 7 |
709 |
|
XM_005273158.1 |
XP_005273215.1
|
6, 7 |
975 |
|
GenBank mRNA |
AK296275.1 |
- |
4, 5 |
242 |
|
AK316272.1 |
- |
2, 3 |
110 |
|
BC013868.2 |
- |
6, 7 |
696 |
|
BC094879.1 |
- |
6, 7 |
658 |
3078971 |
BC117188.1 |
- |
6, 7 |
591 |
40125739 |
BC126173.1 |
- |
6, 7 |
591 |
8991889 |
View Full Product Details