Gene Transcripts
- Gene Symbol
- SNX17
- Entrez Gene ID
- 9784
- Gene Name
- sorting nexin 17
- Gene Aliases
- Chromosome Location
- Chr. 2: 27370496 - 27377533 on Build GRCh38
- UniGene ID
- Hs.278569
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001267059.1 |
NP_001253988.1
|
3, 4 |
473 |
|
NM_001267060.1 |
NP_001253989.1
|
2, 3 |
434 |
|
NM_001267061.1 |
NP_001253990.1
|
3, 4 |
344 |
|
NM_014748.3 |
NP_055563.1
|
3, 4 |
509 |
|
NR_049782.1 |
|
3, 4 |
634 |
|
GenBank mRNA |
AK129689.1 |
- |
3, 4 |
393 |
|
AK222543.1 |
- |
3, 4 |
392 |
|
AK293464.1 |
- |
3, 4 |
325 |
|
AK297054.1 |
- |
3, 4 |
518 |
|
AK298620.1 |
- |
3, 4 |
344 |
|
AK298869.1 |
- |
2, 3 |
295 |
|
AK300144.1 |
- |
3, 4 |
356 |
|
BC002524.1 |
- |
3, 4 |
370 |
3140610 |
BC002610.1 |
- |
3, 4 |
359 |
3143104 |
BC014620.2 |
- |
3, 4 |
370 |
3010648 |
BC050590.1 |
- |
3, 4 |
364 |
6044270 |
BT007167.1 |
- |
3, 4 |
261 |
|
CR457081.1 |
- |
3, 4 |
261 |
|
D31764.1 |
- |
3, 4 |
483 |
|
DB177194.1 |
- |
3, 4 |
499 |
|
View Full Product Details