Gene Transcripts

Gene Symbol
Gdap2
Entrez Gene ID
14547
Gene Name
ganglioside-induced differentiation-associated-protein 2
Gene Aliases
C77050, D3Ertd801e
Chromosome Location
Chr. 3: 100162463 - 100206989 on Build GRCm38
UniGene ID
Mm.279228
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010269.3 NP_034399.1 14 2802
GenBank mRNA AK004887.1 - 14 2748

View Full Product Details