Gene Transcripts

Gene Symbol
Cenpc1
Entrez Gene ID
12617
Gene Name
centromere protein C1
Gene Aliases
CENP-C, Cenpc
Chromosome Location
Chr. 5: 86012018 - 86066019 on Build GRCm38
UniGene ID
Mm.4649
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007683.3 NP_031709.2 6 662
GenBank mRNA AK011004.1 - 6 631
AK137803.1 - 6 631
AK137953.1 - 6 628
AK143566.1 - 6 663
BC150700.1 - 6 458 9087611
U03113.1 - 6 548

View Full Product Details