Gene Transcripts

Gene Symbol
Uba7
Entrez Gene ID
74153
Gene Name
ubiquitin-like modifier activating enzyme 7
Gene Aliases
1300004C08Rik, Ube1l
Chromosome Location
Chr. 9: 107975567 - 107984056 on Build GRCm38
UniGene ID
Mm.277125
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023738.4 NP_076227.1 18 2250
GenBank mRNA AF288694.1 - 13 1512
AK004894.2 - 18 2251
AK054410.1 - 17 2342
AK089235.1 - 17 2143
AK089256.1 - 9 1919
AK129009.1 - 18 2160
AK157941.1 - 1 333
AK170421.1 - 17 2328
AK170644.1 - 16 2709
AK172467.1 - 17 2588
BC037693.1 - 16 2024
BC138843.1 - 18 2167 8861865

View Full Product Details