Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001159941.1 | NP_001153413.1 | 7 | 987 | |
NM_026145.4 | NP_080421.2 | 7 | 984 | ||
GenBank mRNA | AK028657.1 | - | 7 | 951 | |
AK041516.1 | - | 7 | 951 | ||
AK145295.1 | - | 7 | 963 | ||
AK147714.1 | - | 7 | 929 | ||
AK161199.1 | - | 7 | 941 | ||
AK163351.1 | - | 7 | 938 | ||
AK166505.1 | - | 7 | 963 | ||
BC006935.1 | - | 7 | 965 | 3601917 |