Gene Transcripts

Gene Symbol
Hrg
Entrez Gene ID
94175
Gene Name
histidine-rich glycoprotein
Gene Aliases
AI265597, AW413091, D16jh2, D18020, Hprg, Hrgp
Chromosome Location
Chr. 16: 22951072 - 22961659 on Build GRCm38
UniGene ID
Mm.358794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053176.2 NP_444406.2 4 586
GenBank mRNA AB055897.1 - 4 544
AF194028.1 - 4 526
AY135662.1 - 4 547
BC011168.1 - 4 532 4195766
CB593922.1 - 4 558

View Full Product Details