Gene Transcripts
- Gene Symbol
- Scamp4
- Entrez Gene ID
- 56214
- Gene Name
- secretory carrier membrane protein 4
- Gene Aliases
- 2410022D05Rik, AI448940, AU022392, AW742369, Sc4
- Chromosome Location
- Chr. 10: 80602882 - 80615783 on Build GRCm38
- UniGene ID
- Mm.273170
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019575.4 |
NP_062521.1
|
4, 5 |
412 |
|
GenBank mRNA |
AF224721.1 |
- |
4, 5 |
387 |
|
AF295102.1 |
- |
4, 5 |
405 |
|
AK019112.1 |
- |
4, 5 |
415 |
|
AK078226.1 |
- |
4, 5 |
403 |
|
AK154094.1 |
- |
4, 5 |
403 |
|
AK160755.1 |
- |
4, 5 |
403 |
|
AK168554.1 |
- |
4, 5 |
398 |
|
BC018215.1 |
- |
4, 5 |
381 |
3965223 |
BC054104.2 |
- |
4, 5 |
407 |
30021104 |
View Full Product Details