Gene Transcripts

Gene Symbol
Thbs3
Entrez Gene ID
21827
Gene Name
thrombospondin 3
Gene Aliases
TSP3, Thbs-3
Chromosome Location
Chr. 3: 89215142 - 89226837 on Build GRCm38
UniGene ID
Mm.2114
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013691.2 NP_038719.2 4 689
GenBank mRNA BC053023.1 - 4 643
BC060104.1 - 3 1446
L04302.1 - 4 669
L24434.1 - 4 666

View Full Product Details