Gene Transcripts

Gene Symbol
Abi2
Entrez Gene ID
286928
Gene Name
abl-interactor 2
Gene Aliases
Chromosome Location
Chr. 9: 67234612 - 67284504 on Build Rnor_6.0
UniGene ID
Rn.11316
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173143.1 NP_775166.1 2 254
GenBank mRNA BC097273.1 - 312
FQ222350.1 - 624
U94904.1 - 2 254

View Full Product Details