Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_206815.1 | NP_996538.1 | 1 | 308 | |
XM_006251871.2 | XP_006251933.1 | 2 | 377 | ||
XM_006251872.2 | XP_006251934.1 | 2 | 676 | ||
XM_008770517.1 | XP_008768739.1 | 3 | 521 | ||
XM_008770518.1 | XP_008768740.1 | 3 | 495 | ||
XM_008770519.1 | XP_008768741.1 | 2 | 395 | ||
XM_008770520.1 | XP_008768742.1 | 2 | 374 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | XM_008773787.1 | XP_008772009.1 | 2 | 377 | |
XM_008773788.1 | XP_008772010.1 | 2 | 676 | ||
XM_008773789.1 | XP_008772011.1 | 3 | 524 | ||
XM_008773790.1 | XP_008772012.1 | 3 | 498 | ||
XM_008773791.1 | XP_008772013.1 | 2 | 398 | ||
XM_008773792.1 | XP_008772014.1 | 2 | 374 |