Gene Transcripts

Gene Symbol
OR7G3
Entrez Gene ID
390883
Gene Name
olfactory receptor family 7 subfamily G member 3
Gene Aliases
OST085
Chromosome Location
Chr. 19: 9126012 - 9126950 on Build GRCh38
UniGene ID
Hs.553775
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001958.1 NP_001001958.1 1 818

View Full Product Details