Gene Transcripts
- Gene Symbol
- C16orf86
- Entrez Gene ID
- 388284
- Gene Name
- chromosome 16 open reading frame 86
- Gene Aliases
- Chromosome Location
- Chr. 16: 67666806 - 67670523 on Build GRCh38
- UniGene ID
- Hs.632208
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK123796.1 |
- |
2 |
1079 |
|
BC132878.1 |
- |
1 |
644 |
|
BC132880.1 |
- |
1 |
644 |
|
View Full Product Details