Gene Transcripts

Gene Symbol
Rmnd5b
Entrez Gene ID
497900
Gene Name
required for meiotic nuclear division 5 homolog B (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 10: 34979902 - 34990927 on Build Rnor_6.0
UniGene ID
Rn.8249
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017473.1 NP_001017473.1 6 761
XM_006246205.1 XP_006246267.1 6 789
GenBank mRNA BC093386.1 - 6 761

Gene Symbol
LOC103689927
Entrez Gene ID
103689927
Gene Name
protein RMD5 homolog B
Gene Aliases
Chromosome Location
Chr. 10: 37073588 - 37084613 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006246315.1 XP_006246377.1 6 789
XM_008767740.1 XP_008765962.1 6 819

View Full Product Details