Gene Transcripts

Gene Symbol
FEM1A
Entrez Gene ID
55527
Gene Name
fem-1 homolog A
Gene Aliases
EPRAP
Chromosome Location
Chr. 19: 4791716 - 4795559 on Build GRCh38
UniGene ID
Hs.515082
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018708.2 NP_061178.1 1 2112
GenBank mRNA AJ311359.1 - 1 1972
AK074232.1 - 1 2070
AK126532.1 - 2 1840
AK315554.1 - 1 2084
AL359589.1 - 1 250
AL365408.1 - 1 567
AY356352.1 - 1 2066
BC004988.1 - 1 2070 2957950
CR749732.1 - 1 250

View Full Product Details