Gene Transcripts

Gene Symbol
SPRED3
Entrez Gene ID
399473
Gene Name
sprouty related EVH1 domain containing 3
Gene Aliases
Eve-3, spred-3
Chromosome Location
Chr. 19: 38390061 - 38399884 on Build GRCh38
UniGene ID
Hs.196011
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042522.2 NP_001035987.1 2 339
NR_073032.1 2 154
XM_006723219.3 XP_006723282.1 3 330
XM_011526978.2 XP_011525280.1 2 385
GenBank mRNA BC151136.1 - 2 339
BF983829.1 - 2 156
DQ323927.1 - 2 240
DQ323928.1 - 2 240

View Full Product Details