Gene Transcripts
- Gene Symbol
- LINC00521
- Entrez Gene ID
- 256369
- Gene Name
- long intergenic non-protein coding RNA 521
- Gene Aliases
- C14orf48, c14_5713
- Chromosome Location
- Chr. 14: 93997270 - 94011695 on Build GRCh38
- UniGene ID
- Hs.143845
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024184.1 |
|
6, 7 |
1028 |
|
GenBank mRNA |
BC021728.2 |
- |
6, 7 |
1027 |
|
View Full Product Details