Gene Transcripts
- Gene Symbol
- SYNE4
- Entrez Gene ID
- 163183
- Gene Name
- spectrin repeat containing nuclear envelope family member 4
- Gene Aliases
- C19orf46, DFNB76, Nesp4
- Chromosome Location
- Chr. 19: 36003299 - 36008793 on Build GRCh38
- UniGene ID
- Hs.436743
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK093764.1 |
- |
5 |
1625 |
|
View Full Product Details