Gene Transcripts

Gene Symbol
DIABLO
Entrez Gene ID
56616
Gene Name
diablo IAP-binding mitochondrial protein
Gene Aliases
DFNA64, SMAC
Chromosome Location
Chr. 12: 122207662 - 122227534 on Build GRCh38
UniGene ID
Hs.169611
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278302.1 NP_001265231.1 3 780
NM_001278303.1 NP_001265232.1 3 808
NM_001278304.1 NP_001265233.1 3 1410
NM_001278342.1 NP_001265271.1 3 809
NM_019887.5 NP_063940.1 5 1205
NM_138930.3 NP_620308.1 4 912
GenBank mRNA AF262240.1 - 4 406
AF298770.1 - 3 228
AK001399.1 - 3 1414
AK024768.1 - 4 414
AK057778.1 - 3 282
AK222664.1 - 4 403
AK315629.1 - 4 425
AY313210.1 - 3 255
BC004417.1 - 2 220
BC011909.2 - 4 410 4137792
BC046209.1 - 5 1127
BC095468.1 - 4 396 30557745
BF313712.1 - 3 282
BF724951.1 - 3 276
BM746689.1 - 3 251
CF128595.1 - 4 396
JX679472.1 - 2 96
KU178725.1 - 4 387
KU178726.1 - 3 255

View Full Product Details