Gene Transcripts
- Gene Symbol
- EVA1B
- Entrez Gene ID
- 55194
- Gene Name
- eva-1 homolog B (C. elegans)
- Gene Aliases
- C1orf78, FAM176B
- Chromosome Location
- Chr. 1: 36322030 - 36324154 on Build GRCh38
- UniGene ID
- Hs.87016
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK001509.1 |
- |
3 |
378 |
|
View Full Product Details